Learn More
Medchemexpress LLC HY-130603 5mg Medchemexpress, DCZ0415 CAS:2242470-43-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1306035MG
Encompass_Preferred
Medchemexpress, HY-130603 5mg DCZ0415 CAS:2242470-43-3 DCZ0415, a potent TRIP13 inhibitor, impairs nonhomologous end joining repair and inhibits NF-κB activity. DCZ0415 induces anti-myeloma activity in vitro, in vivo, and in primary cells derived from drug-resistant myeloma patients. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0211