Learn More
Medchemexpress LLC DD-03-156 | 2769753-69-5 | 99.9% | 1106.31 | 100 MG
SDP
Catalog No. 5000352157
Encompass_Preferred
DD-03-156 is a potent and selective degrader of CDK17 and LIMK2. Its exquisite selectivity and potency make it an advanced starting point for the development of a chemical probe for the degradation of CDK17. It includes a VHL ligand, a linker, and a BRAF inhibitor.
  • Potent and selective degrader of CDK17 and LIMK2
  • Exquisite selectivity and potency
  • Advanced starting point for developing a chemical probe for CDK17 degradation
  • For research use only
Catalog No. 50-003-52157 Supplier Medchemexpress LLC Supplier No. HY137346100MG
DD-03-156 is a potent and selective degrader of CDK17 and LIMK2. Its exquisite selectivity and potency make it an advanced starting point for the development of a chemical probe for the degradation of CDK17. It includes a VHL ligand, a linker, and a BRAF inhibitor.
  • Potent and selective degrader of CDK17 and LIMK2
  • Exquisite selectivity and potency
  • Advanced starting point for developing a chemical probe for CDK17 degradation
  • For research use only