Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DD-03-171 | 2366132-45-6 | 98.7% | 1 ML
SDP

Catalog No. 5000232367
Encompass_Preferred

PROTAC BTK degrader supplied as a concentrated solution for use in biochemical and cellular research. Provided as a small-volume, high-concentration stock suitable for in vitro assays; reported purity supports use in assay development and preclinical studies.

  • Provided as a 10 mM solution in DMSO, 1 mL.
  • Induces selective degradation of Bruton's tyrosine kinase (BTK) in cellular models.
  • High reported purity, suitable for biochemical and cell-based assays.
  • Chemical formula C55H62N10O8 and molecular weight 991.14 g/mol.
  • Packaged for research applications requiring small-volume, concentrated stocks.

Catalog No. 50-002-32367 Supplier Medchemexpress LLC Supplier No. HY12283010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC BTK degrader supplied as a concentrated solution for use in biochemical and cellular research. Provided as a small-volume, high-concentration stock suitable for in vitro assays; reported purity supports use in assay development and preclinical studies.

  • Provided as a 10 mM solution in DMSO, 1 mL.
  • Induces selective degradation of Bruton's tyrosine kinase (BTK) in cellular models.
  • High reported purity, suitable for biochemical and cell-based assays.
  • Chemical formula C55H62N10O8 and molecular weight 991.14 g/mol.
  • Packaged for research applications requiring small-volume, concentrated stocks.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.