Learn More
Medchemexpress LLC DD-03-171 | 2366132-45-6 | 98.7% | 5 MG
SDP
Catalog No. 5000230763
Encompass_Preferred
A PROTAC small-molecule degrader of Bruton's tyrosine kinase (BTK) intended for research use; degrades BTK, IKZF1, and IKZF3, and inhibits mantle cell lymphoma cell proliferation in reported assays.
  • Degrades BTK, IKZF1, and IKZF3.
  • Reported IC50 approximately 5.1 nM for inhibition of MCL cell proliferation.
  • Available as solid (5 mg-500 mg) and as 10 mM solution in DMSO.
  • High purity (98.7%).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-30763 Supplier Medchemexpress LLC Supplier No. HY1228305MG
A PROTAC small-molecule degrader of Bruton's tyrosine kinase (BTK) intended for research use; degrades BTK, IKZF1, and IKZF3, and inhibits mantle cell lymphoma cell proliferation in reported assays.
  • Degrades BTK, IKZF1, and IKZF3.
  • Reported IC50 approximately 5.1 nM for inhibition of MCL cell proliferation.
  • Available as solid (5 mg-500 mg) and as 10 mM solution in DMSO.
  • High purity (98.7%).
  • For research use only; not for human or clinical use.