Learn More
Aviva Systems Biology Corp DDAH2 Antibody - N-terminal region (ARP58611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDAH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDAH2. Alternate names: DDAH, DDAHII, G6a, NG30ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62374