Learn More
Aat Bioquest DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] *CAS 118290-05-4*
SDP
Catalog No. 501952689
Encompass_Preferred
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] is used as a calibration standard for DDAO-based enzyme substrates. It has pH-dependent red fluorescence with excitation close to the He-Ne red laser line 633 nm.
Catalog No. 50-195-2689 Supplier Aat Bioquest Supplier No. 62
DDAO [7-hydroxy-9H-(1,3-dichloro-9,9-dimethylacridin-2-one)] is used as a calibration standard for DDAO-based enzyme substrates. It has pH-dependent red fluorescence with excitation close to the He-Ne red laser line 633 nm.