Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DDB1 protein, human (sf9, His) | 96.0% | 10UG
SDP

Catalog No. 5000176269
Encompass_Preferred

DDB1 protein is a recombinant human DNA damage-binding protein 1 produced in Sf9 insect cells and supplied as an N-terminal His-tagged, 0.22 μm-filtered solution formulated for biochemical and cell-based research. The preparation is characterized for concentration, purity, molecular weight, endotoxin, and storage stability to support studies of DNA repair, ubiquitination, and protein-protein interactions.

  • Recombinant human DDB1 expressed in Sf9 insect cells.
  • N-terminal His tag for affinity purification and detection.
  • Supplied in 50 mM HEPES, pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • High purity (>90% by reducing SDS-PAGE) suitable for biochemical assays.
  • Provided at 1.45 mg/mL to facilitate assay setup.
  • Low endotoxin level (<1 EU/μg) for sensitive applications.
  • Store at -80°C; stable short term at -20°C; avoid repeated freeze-thaw cycles.

Catalog No. 50-001-76269 Supplier Medchemexpress LLC Supplier No. HYP70356310UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DDB1 protein is a recombinant human DNA damage-binding protein 1 produced in Sf9 insect cells and supplied as an N-terminal His-tagged, 0.22 μm-filtered solution formulated for biochemical and cell-based research. The preparation is characterized for concentration, purity, molecular weight, endotoxin, and storage stability to support studies of DNA repair, ubiquitination, and protein-protein interactions.

  • Recombinant human DDB1 expressed in Sf9 insect cells.
  • N-terminal His tag for affinity purification and detection.
  • Supplied in 50 mM HEPES, pH 7.5, 200 mM NaCl, 20% glycerol, 1 mM DTT.
  • High purity (>90% by reducing SDS-PAGE) suitable for biochemical assays.
  • Provided at 1.45 mg/mL to facilitate assay setup.
  • Low endotoxin level (<1 EU/μg) for sensitive applications.
  • Store at -80°C; stable short term at -20°C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.