Learn More
Sigma Aldrich Fine Chemicals Biosciences DDC Mass Spect GoldR Serum
Catalog No. 502904298
Encompass
DDC Mass Spect Gold is processed from human plasma using a validated process that reduces the hormone and steroid levels please see average hormone and steroid values below. This process also maintains the inherent characteristics of the human plasma resulting a in a pool that is consistent with patient samples. The human plasma is collected from donors at fDA licensed facilities. Each donor is screened for donor eligibility according to fDA regulations. Each unit is tested for and found negative for HIV1/2-ab HBsAg HCV-ab HIV-1 (NAT) HCV (NAT) and RPR. The plasma is inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records raw material inspection records batch records testing
Catalog No. 50-290-4298 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MSG3000100ML
DDC Mass Spect Gold is processed from human plasma using a validated process that reduces the hormone and steroid levels please see average hormone and steroid values below. This process also maintains the inherent characteristics of the human plasma resulting a in a pool that is consistent with patient samples. The human plasma is collected from donors at fDA licensed facilities. Each donor is screened for donor eligibility according to fDA regulations. Each unit is tested for and found negative for HIV1/2-ab HBsAg HCV-ab HIV-1 (NAT) HCV (NAT) and RPR. The plasma is inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records raw material inspection records batch records testing