Learn More
Medchemexpress LLC ddCTP (trisodium) solution (100mM) | 98.0% | 517.10 | 2.58 MG
SDP
Catalog No. 5000352188
Encompass_Preferred
ddCTP trisodium is one of 2',3'-dideoxyribonucleoside 5'-triphosphates (ddNTPs) that acts as a chain-elongating inhibitor of DNA polymerase for DNA sequencing. It is a nucleoside analog that targets the reverse transcriptase of human immunodeficiency virus (HIV) and can be used for AIDS research.
  • Functions as a chain-elongating inhibitor of DNA polymerase for DNA sequencing.
  • Targets the reverse transcriptase of human immunodeficiency virus (HIV).
  • Suitable for AIDS research.
  • Acts as a nucleoside analog.
Catalog No. 50-003-52188 Supplier Medchemexpress LLC Supplier No. HY137697D2.58MG
ddCTP trisodium is one of 2',3'-dideoxyribonucleoside 5'-triphosphates (ddNTPs) that acts as a chain-elongating inhibitor of DNA polymerase for DNA sequencing. It is a nucleoside analog that targets the reverse transcriptase of human immunodeficiency virus (HIV) and can be used for AIDS research.
  • Functions as a chain-elongating inhibitor of DNA polymerase for DNA sequencing.
  • Targets the reverse transcriptase of human immunodeficiency virus (HIV).
  • Suitable for AIDS research.
  • Acts as a nucleoside analog.