Learn More
TARGETMOL CHEMICALS INC DDD100097 5MG
SDP
Catalog No. 502580837
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DDD100097 is a potent N-myristoyltransferase inhibitor with potential inhibitory effects on Trypanosoma brucei which can be used to study neurological disorders. purity: 99%
Catalog No. 50-258-0837 Supplier TARGETMOL CHEMICALS INC Supplier No. T239735MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. DDD100097 is a potent N-myristoyltransferase inhibitor with potential inhibitory effects on Trypanosoma brucei which can be used to study neurological disorders. purity: 99%