Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Butanedioic acid, compd. with cabamiquine (6-fluoro quinoline carboxamide) | 2444781-71-7 | 99.8% | 580.65 | C31H37FN4O6 | 10 MG
SDP

Catalog No. 501969489
Encompass_Preferred

Cabamiquine succinate is the succinate salt of cabamiquine (DDD107498), a potent orally active antimalarial research compound that inhibits Plasmodium protein synthesis via eEF2/CaMKIII. It is supplied as a white to off-white solid with high analytical purity and intended for laboratory research.

  • High purity: 99.8% (LCMS).
  • Suitable for in vitro and preclinical assays targeting Plasmodium species.
  • Soluble in DMSO (≈135 mg/mL); ultrasonic agitation may be required.
  • Storage: sealed, away from moisture and light; in solvent store -80°C (6 months) or -20°C (1 month).
  • Available in small mg-scale packages for research applications.

Catalog No. 50-196-9489 Supplier Medchemexpress LLC Supplier No. HY117684A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cabamiquine succinate is the succinate salt of cabamiquine (DDD107498), a potent orally active antimalarial research compound that inhibits Plasmodium protein synthesis via eEF2/CaMKIII. It is supplied as a white to off-white solid with high analytical purity and intended for laboratory research.

  • High purity: 99.8% (LCMS).
  • Suitable for in vitro and preclinical assays targeting Plasmodium species.
  • Soluble in DMSO (≈135 mg/mL); ultrasonic agitation may be required.
  • Storage: sealed, away from moisture and light; in solvent store -80°C (6 months) or -20°C (1 month).
  • Available in small mg-scale packages for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.