Learn More
Medchemexpress LLC DDD85646 | 1215010-55-1 | 99.7% | 495.43 g/mol | C21H24Cl2N6O2S | 10 MG
SDP
Catalog No. 5000246789
Encompass_Preferred
DDD85646 (IMP-366) is a potent N-myristoyltransferase (NMT) inhibitor used in anti-trypanosomal research. Supplied as a high-purity powder, it is suitable for in vitro and in vivo studies and includes manufacturer-recommended formulation protocols for animal dosing.
  • Potent NMT inhibitor active against Trypanosoma brucei.
  • High purity (99.72%) suitable for biochemical and cell assays.
  • Soluble in DMSO at 100 mg/mL; in vivo formulation protocols provided.
  • Supplied as powder in research-scale quantities for laboratory studies.
  • Accompanied by an SDS containing molecular formula and molecular weight details.
Catalog No. 50-002-46789 Supplier Medchemexpress LLC Supplier No. HY10305610MG
DDD85646 (IMP-366) is a potent N-myristoyltransferase (NMT) inhibitor used in anti-trypanosomal research. Supplied as a high-purity powder, it is suitable for in vitro and in vivo studies and includes manufacturer-recommended formulation protocols for animal dosing.
  • Potent NMT inhibitor active against Trypanosoma brucei.
  • High purity (99.72%) suitable for biochemical and cell assays.
  • Soluble in DMSO at 100 mg/mL; in vivo formulation protocols provided.
  • Supplied as powder in research-scale quantities for laboratory studies.
  • Accompanied by an SDS containing molecular formula and molecular weight details.