Learn More
Medchemexpress LLC DDD85646 (IMP-366) | 1215010-55-1 | 99.7% | 495.43 g/mol | C21H24Cl2N6O2S | 1 MG
SDP
Catalog No. 5000247625
Encompass_Preferred
DDD85646 (IMP-366) is a research-grade small-molecule inhibitor of N-myristoyltransferase (NMT) with low-nanomolar activity against Trypanosoma brucei and human NMT. It is supplied with purity documentation and is intended for laboratory research use only.
  • Low-nanomolar potency against TbNMT and hNMT (reported IC50s ~2 nM and ~4 nM)
  • Available as solid or 10 mM solutions in DMSO for immediate use
  • High reported purity (≈99.7%) with accompanying documentation
  • Provided in multiple solid package sizes to support screening and scale-up
  • Suitable for biochemical assays and cell-based NMT research
Catalog No. 50-002-47625 Supplier Medchemexpress LLC Supplier No. HY1030561MG
DDD85646 (IMP-366) is a research-grade small-molecule inhibitor of N-myristoyltransferase (NMT) with low-nanomolar activity against Trypanosoma brucei and human NMT. It is supplied with purity documentation and is intended for laboratory research use only.
  • Low-nanomolar potency against TbNMT and hNMT (reported IC50s ~2 nM and ~4 nM)
  • Available as solid or 10 mM solutions in DMSO for immediate use
  • High reported purity (≈99.7%) with accompanying documentation
  • Provided in multiple solid package sizes to support screening and scale-up
  • Suitable for biochemical assays and cell-based NMT research