Learn More
Medchemexpress LLC Dde Biotin-PEG4-alkyne | 1802908-00-4 | 99.3% | C32H50N4O8S | 100 MG
SDP
Catalog No. 5000354897
Encompass_Preferred
Dde Biotin-PEG4-alkyne is a polyethylene glycol (PEG)-based PROTAC linker designed for use in the synthesis of PROTACs (Proteolysis Targeting Chimeras). This compound also functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. PROTACs are bifunctional molecules that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein via a linker.
  • It is a peg-based protac linker.
  • It contains an alkyne group.
  • It is suitable for copper-catalyzed azide-alkyne cycloaddition (cuaac) with azide-containing molecules.
  • It is intended for research use only.
Catalog No. 50-003-54897 Supplier Medchemexpress LLC Supplier No. HY140924100MG
Dde Biotin-PEG4-alkyne is a polyethylene glycol (PEG)-based PROTAC linker designed for use in the synthesis of PROTACs (Proteolysis Targeting Chimeras). This compound also functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. PROTACs are bifunctional molecules that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein via a linker.
  • It is a peg-based protac linker.
  • It contains an alkyne group.
  • It is suitable for copper-catalyzed azide-alkyne cycloaddition (cuaac) with azide-containing molecules.
  • It is intended for research use only.