Learn More
Medchemexpress LLC Dde Biotin-PEG4-alkyne | 1802908-00-4 | 99.3% | C32H50N4O8S | 250 MG
SDP
Catalog No. 5000352730
Encompass_Preferred
Dde Biotin-PEG4-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Possesses an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables reaction with molecules containing azide groups
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system
Catalog No. 50-003-52730 Supplier Medchemexpress LLC Supplier No. 250MGHY140924
Dde Biotin-PEG4-alkyne is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Possesses an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables reaction with molecules containing azide groups
  • Facilitates targeted protein degradation via the ubiquitin-proteasome system