Learn More
Medchemexpress LLC Dde biotin-PEG4-azide | 1802907-93-2 | 99.4% | C32H53N7O8S | 10MG
SDP
Catalog No. 5000260468
Encompass_Preferred
Dde Biotin-PEG4-azide is a biotinylated PEG4 azide reagent for introducing a biotin tag into alkyne-containing biomolecules via Cu(I)-catalyzed azide-alkyne cycloaddition (click chemistry). It contains a removable Dde protecting group and a PEG4 spacer to improve solubility and provide defined linker length for probe construction and conjugation workflows.
  • Provides a biotin handle for affinity capture and detection.
  • Contains a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Features an azide functional group for copper-catalyzed click chemistry with alkynes.
  • Includes a Dde removable protecting group for orthogonal deprotection.
  • Supplied at high chemical purity for reliable conjugation results.
  • Suitable for protein labeling, probe synthesis, and linker construction.
Catalog No. 50-002-60468 Supplier Medchemexpress LLC Supplier No. HY14091610MG
Dde Biotin-PEG4-azide is a biotinylated PEG4 azide reagent for introducing a biotin tag into alkyne-containing biomolecules via Cu(I)-catalyzed azide-alkyne cycloaddition (click chemistry). It contains a removable Dde protecting group and a PEG4 spacer to improve solubility and provide defined linker length for probe construction and conjugation workflows.
  • Provides a biotin handle for affinity capture and detection.
  • Contains a PEG4 spacer to improve solubility and reduce steric hindrance.
  • Features an azide functional group for copper-catalyzed click chemistry with alkynes.
  • Includes a Dde removable protecting group for orthogonal deprotection.
  • Supplied at high chemical purity for reliable conjugation results.
  • Suitable for protein labeling, probe synthesis, and linker construction.