Learn More
Medchemexpress LLC Dde Biotin-PEG4-DBCO | 1807512-43-1 | 96.2% | C47H61N5O9S | 25 MG
SDP
Catalog No. 5000357375
Encompass_Preferred
Dde Biotin-PEG4-DBCO is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains a DBCO group that undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules
Catalog No. 50-003-57375 Supplier Medchemexpress LLC Supplier No. 25MGHY140931
Dde Biotin-PEG4-DBCO is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains a DBCO group that undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with azide-containing molecules