Learn More
Medchemexpress LLC Dde Biotin-PEG4-DBCO | 1807512-43-1 | 96.2% | C47H61N5O9S | 50 MG
SDP
Catalog No. 5000353169
Encompass_Preferred
Dde Biotin-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules
Catalog No. 50-003-53169 Supplier Medchemexpress LLC Supplier No. HY14093150MG
Dde Biotin-PEG4-DBCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules