Learn More
New England Biolabs, Inc. DdeI – 1000 units
SDP
Catalog No. 50812089
Encompass
A restriction endonuclease that recognizes the sequence C^TNA_G.
Catalog No. 50-812-089 Supplier New England Biolabs, Inc. Supplier No. R0175S
A restriction endonuclease that recognizes the sequence C^TNA_G.