Learn More
New England Biolabs, Inc. DdeI – 5000 units
SDP
Catalog No. 50812088
Encompass
A restriction endonuclease that recognizes the sequence C^TNA_G.
Catalog No. 50-812-088 Supplier New England Biolabs, Inc. Supplier No. R0175L
A restriction endonuclease that recognizes the sequence C^TNA_G.