Learn More
American Radiolabeled Chemicals Inc DDG 14C(U) D 50UC
SDP
Catalog No. 50352930
Encompass_Preferred
Deoxy-D-glucose, 2-14C(U) 50uCi in ethanol:water (3:97), 0.1 mCi/ml, 300-360 mCi/mmol, M.W. 164.2
Catalog No. 50-352-930 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 0112D50UC
Deoxy-D-glucose, 2-14C(U) 50uCi in ethanol:water (3:97), 0.1 mCi/ml, 300-360 mCi/mmol, M.W. 164.2