Learn More
American Radiolabeled Chemicals Inc DDG 2-1 2-3H A 1LC
SDP
Catalog No. 5090107632
Encompass_Preferred
Deoxy-D-glucose, 2-1,2-3H 1mCi in sterile saline, 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 164.2
Catalog No. 50-901-07632 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 0103A1MC
Deoxy-D-glucose, 2-1,2-3H 1mCi in sterile saline, 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 164.2