Learn More
Medchemexpress LLC ddGTP (trisodium) | 99.7% | 557.13 | 2.78MG
SDP
Supplier:  Medchemexpress LLC 2.78MGHY134103A
Encompass_Preferred
ddGTP (2',3'-Dideoxyguanosine 5'-triphosphate) trisodium is a dideoxyribonucleoside 5'-triphosphate (ddNTP) that acts as a chain-elongating inhibitor of DNA polymerase for DNA sequencing. It also functions as an inhibitor or a substrate for DNA polymerase α.
  • Acts as a chain-elongating inhibitor of DNA polymerase
  • Used for DNA sequencing applications
  • Functions as an inhibitor or substrate for DNA polymerase α
  • Inhibits the formation of both negative and positive DNA strands
  • Competitively inhibits the incorporation of [3H]dGTP into DNA
Catalog No. 50-003-66082