Learn More
Aviva Systems Biology Corp DDI1 Antibody - middle region (ARP56142_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56142_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DDI1. Alternate names: FLJ36017ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61736