Learn More
OriGene DDKTrap Anti-DDK Affinity Resin, 5 mL
SDP
Catalog No. 5000600182
Encompass
DDKTrap Anti-DDK 8G6 affinity resin used for the purification of DDK, DYKDDDDK, or FLAG tagged proteins , 5 mLApplications: Affinity Chromatography, IP, Protein purificationBuffer: 50% glycerol, 1 x PBS, 0.02% NaN3Binding Ligand: Anti-DDK Antibody, Clone 8G6Matrix: 4% agarose microspheresTarget Protein Binding Capacity: >1 mg DDK-tagged protein/ml resinPurity: >95% conjugated antibody purity determined by Coomassie Blue stainingStability: Stable for 12 months from the date of receipt of the product under proper storage and handing conditions. Avoid repeat exposures to room temperature.Storage: Store at -20°C immediately upon receipt.
Catalog No. 50-006-00182 Supplier OriGene Supplier No. AR100028M
DDKTrap Anti-DDK 8G6 affinity resin used for the purification of DDK, DYKDDDDK, or FLAG tagged proteins , 5 mLApplications: Affinity Chromatography, IP, Protein purificationBuffer: 50% glycerol, 1 x PBS, 0.02% NaN3Binding Ligand: Anti-DDK Antibody, Clone 8G6Matrix: 4% agarose microspheresTarget Protein Binding Capacity: >1 mg DDK-tagged protein/ml resinPurity: >95% conjugated antibody purity determined by Coomassie Blue stainingStability: Stable for 12 months from the date of receipt of the product under proper storage and handing conditions. Avoid repeat exposures to room temperature.Storage: Store at -20°C immediately upon receipt.