Learn More
Medchemexpress LLC DDL-357 | 3077900-08-1 | 98.3% | 440.95 | 100 MG
SDP
Catalog No. 5000386974
Encompass_Preferred
DDL-357 is a potent secreted clusterin enhancer that reduces phospho-tau in the brain and improves memory in the murine 3xTg-AD model. It is for research use only and not sold to patients.
  • Potent secreted clusterin enhancer
  • Reduces phospho-tau in brain
  • Improves memory in the murine 3xTg-AD model
  • For research use only
Catalog No. 50-003-86974 Supplier Medchemexpress LLC Supplier No. HY176438100MG
DDL-357 is a potent secreted clusterin enhancer that reduces phospho-tau in the brain and improves memory in the murine 3xTg-AD model. It is for research use only and not sold to patients.
  • Potent secreted clusterin enhancer
  • Reduces phospho-tau in brain
  • Improves memory in the murine 3xTg-AD model
  • For research use only