Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DDM/CHS Solution (10:1)

Catalog No. FERA50942 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA50942 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA50942 Supplier Thermo Scientific™ Supplier No. A50942
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A ready-to-use solution for solubilization of membrane proteins while maintaining structural integrity and activity.

  • A ready-to-use solution for solubilization of membrane proteins while maintaining structural integrity and activity
  • The stable pre-formulated solution contains a mixture of dodecyl-β-D-maltoside (DDM) (100 mg/mL) and cholesteryl hemisuccinate (CHS) (10 mg/mL) in deionized water with final concentrations of 10% and 1%, respectively
  • Typical working concentrations are 1% DDM/0.1% CHS for protein extraction and 0.1% DDM/0.01% CHS for protein purification

Features of DDM/CHS Solution include:

  • Ready-to-use liquid formulation
  • Mixture of nonionic detergent, DDM, with dual hydrophobic/hydrophilic properties and anionic detergent, CHS, that aids in the stabilization of large unilamellar vesicles
  • DDM/CHS micelles aid in solubilization of membrane proteins while preserving structure and function
  • Suitable for protein solubilization and stabilization for single particle cryo-EM analysis

Properties of DDM:

  • Molecular formula: C24H46O11
  • Molecular weight: 510.6 g/mol
  • Detergent class: Non-ionic
  • Micelle molecular weight: 50,000 g
  • Critical micelle concentration (CMC): 0.17 mM (0.009% w/v) in water; 0.12 mM (0.006% w/v) in 0.2 M NaCl

Properties of CHS:

  • Molecular formula: C31H50O4 · C4H11NO3
  • Molecular weight: 607.9 g/mol
  • Detergent: Ionizable anionic
  • Function: Stabilizes large unilamellar vesicles

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Quantity 10 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.