Learn More
Medchemexpress LLC DDO-2213 | 2378768-36-4 | C24H27ClFN7O | 100 MG
SDP
Catalog No. 5000373755
Encompass_Preferred
DDO-2213 is an orally active and potent WDR5-MLL1 inhibitor. It selectively inhibits MLL histone methyltransferase activity and the proliferation of MLL translocation-harboring cells.
  • Orally active and potent WDR5-MLL1 inhibitor
  • Selectively inhibits MLL histone methyltransferase activity
  • Inhibits proliferation of MLL translocation-harboring cells
  • Suitable for MLL fusion leukemia research
Catalog No. 50-003-73755 Supplier Medchemexpress LLC Supplier No. HY153331100MG
Add to Cart
Add to Cart
DDO-2213 is an orally active and potent WDR5-MLL1 inhibitor. It selectively inhibits MLL histone methyltransferase activity and the proliferation of MLL translocation-harboring cells.
  • Orally active and potent WDR5-MLL1 inhibitor
  • Selectively inhibits MLL histone methyltransferase activity
  • Inhibits proliferation of MLL translocation-harboring cells
  • Suitable for MLL fusion leukemia research