Learn More
Medchemexpress LLC DDO-2213 | 2378768-36-4 | C24H27ClFN7O | 5 MG
SDP
Catalog No. 5000372590
Encompass_Preferred
DDO-2213 is an orally active and potent WDR5-MLL1 inhibitor, with an IC50 of 29 nM and a Kd value of 72.9 nM for the WDR5 protein. It selectively inhibits MLL (mixed lineage leukemia) histone methyltransferase activity and the proliferation of MLL translocation-harboring cells. DDO-2213 can be used for MLL fusion leukemia research.
  • Orally active
  • Potent WDR5-MLL1 inhibitor
  • Selectively inhibits MLL histone methyltransferase activity
  • Inhibits proliferation of MLL translocation-harboring cells
  • Useful for MLL fusion leukemia research
Catalog No. 50-003-72590 Supplier Medchemexpress LLC Supplier No. HY1533315MG
DDO-2213 is an orally active and potent WDR5-MLL1 inhibitor, with an IC50 of 29 nM and a Kd value of 72.9 nM for the WDR5 protein. It selectively inhibits MLL (mixed lineage leukemia) histone methyltransferase activity and the proliferation of MLL translocation-harboring cells. DDO-2213 can be used for MLL fusion leukemia research.
  • Orally active
  • Potent WDR5-MLL1 inhibitor
  • Selectively inhibits MLL histone methyltransferase activity
  • Inhibits proliferation of MLL translocation-harboring cells
  • Useful for MLL fusion leukemia research