Learn More
Medchemexpress LLC Ddo-5936 | 2355377-13-6 | MFCD33398564 | 99.2% | 495.59 g·mol⁻¹ | C25H29N5O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1393015MG
Encompass_Preferred
DDO-5936 is a small-molecule inhibitor that selectively disrupts the protein-protein interaction between heat shock protein 90 (Hsp90) and Cdc37. It is used in preclinical research to probe Hsp90-Cdc37-dependent signaling and has been applied in colorectal cancer studies. Supplied as a high-purity solid for solution preparation and biological testing.
  • Potent and specific Hsp90-Cdc37 PPI inhibitor.
  • High purity (99.2%) suitable for biochemical assays.
  • Provided as a solid for stable storage and handling.
  • Soluble in common polar organic solvents for assay preparation.
  • Validated in cellular and in vivo colorectal cancer models.
Catalog No. 50-002-21152