Learn More
TARGETMOL CHEMICALS INC DDO-7263 5MG
SDP
Catalog No. 502579615
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DDO-7263 is a 124-Oxadiazole derivative that upregulates Nrf2 through binding to Rpn6 to block the assembly of 26S proteasome and the subsequent degradation of ubiquitinated Nrf2. DDO-7263 is a potent Nrf2 activator that activates the Nrf2-ARE signaling pathway and exerts anti-inflammatory activity [1]. purity: 99%
Catalog No. 50-257-9615 Supplier TARGETMOL CHEMICALS INC Supplier No. T604875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DDO-7263 is a 124-Oxadiazole derivative that upregulates Nrf2 through binding to Rpn6 to block the assembly of 26S proteasome and the subsequent degradation of ubiquitinated Nrf2. DDO-7263 is a potent Nrf2 activator that activates the Nrf2-ARE signaling pathway and exerts anti-inflammatory activity [1]. purity: 99%