Learn More
Aviva Systems Biology Corp DDO Antibody - N-terminal region (ARP61286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDO. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DDO. Alternate names: DASOX, DDO-1, DDO-2, FLJ45203ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-287-328