Learn More
Aviva Systems Biology Corp DDOST Antibody - N-terminal region (ARP46585_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46585_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDOST. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDOST. Alternate names: AGE-R1, KIAA0115, MGC2191, OK/SW-cl.45, OST, OST48, WBP1, AGER1, OKSWcl45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59355