Learn More
Medchemexpress LLC DDP-38003 dihydrochloride | 1831167-98-6 | 99.3% | 423.38 | 100 MG
SDP
Catalog No. 5000400491
Encompass_Preferred
DDP-38003 dihydrochloride is a novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM. It exhibits in vivo efficacy after oral administration, determining a 62% increased survival in mouse leukemia models with evidence of KDM1A inhibition. The half-life of DDP-38003 is 8 hours. A significant dose-dependent increase of mice survival is obtained by DDP-38003 treatment. DDP-38003 is a potential oral anticancer agent.
  • Novel, orally available inhibitor of KDM1A/LSD1
  • IC50 of 84 nM against KDM1A
  • More active in reducing colony forming ability and inducing differentiation of THP-1 cells compared to the 1R, 2S analogue
  • Exhibits in vivo efficacy after oral administration
  • Determines 62% increased survival in mouse leukemia models
  • Half-life of 8 hours
  • Potential oral anticancer agent
  • Appearance: Solid, brown to reddish brown
Catalog No. 50-004-00491 Supplier Medchemexpress LLC Supplier No. HY19612A100MG
DDP-38003 dihydrochloride is a novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM. It exhibits in vivo efficacy after oral administration, determining a 62% increased survival in mouse leukemia models with evidence of KDM1A inhibition. The half-life of DDP-38003 is 8 hours. A significant dose-dependent increase of mice survival is obtained by DDP-38003 treatment. DDP-38003 is a potential oral anticancer agent.
  • Novel, orally available inhibitor of KDM1A/LSD1
  • IC50 of 84 nM against KDM1A
  • More active in reducing colony forming ability and inducing differentiation of THP-1 cells compared to the 1R, 2S analogue
  • Exhibits in vivo efficacy after oral administration
  • Determines 62% increased survival in mouse leukemia models
  • Half-life of 8 hours
  • Potential oral anticancer agent
  • Appearance: Solid, brown to reddish brown