Learn More
Medchemexpress LLC DDP-38003 dihydrochloride | 1831167-98-6 | 99.3% | 423.38 | 50 MG
SDP
Catalog No. 5000401268
Encompass_Preferred
DDP-38003 dihydrochloride is a novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM. It demonstrates significant in vitro and in vivo efficacy, inhibiting KDM1A and reducing colony forming ability in THP-1 cells. In mouse leukemia models, oral administration of DDP-38003 dihydrochloride led to increased survival.
  • Inhibits histone lysine-specific demethylase 1A (KDM1A/LSD1)
  • Orally available
  • IC50 of 84 nM for KDM1A
  • Reduces colony forming ability and induces differentiation of THP-1 cells
  • Shows in vivo efficacy in mouse leukemia models
  • Potential oral anticancer agent
Catalog No. 50-004-01268 Supplier Medchemexpress LLC Supplier No. HY19612A50MG
DDP-38003 dihydrochloride is a novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM. It demonstrates significant in vitro and in vivo efficacy, inhibiting KDM1A and reducing colony forming ability in THP-1 cells. In mouse leukemia models, oral administration of DDP-38003 dihydrochloride led to increased survival.
  • Inhibits histone lysine-specific demethylase 1A (KDM1A/LSD1)
  • Orally available
  • IC50 of 84 nM for KDM1A
  • Reduces colony forming ability and induces differentiation of THP-1 cells
  • Shows in vivo efficacy in mouse leukemia models
  • Potential oral anticancer agent