Learn More
Bio Rad Laboratories ddPCR Buffer Control for Probes
Catalog No. NC1228187
Encompass
9 mL (2 x 4.5 mL), 2x buffer, for use as blank control with Droplet Generation Oil for Probes.
This item is not returnable. View return policy
Catalog No. NC1228187 Supplier Bio Rad Laboratories Supplier No. 1863052
9 mL (2 x 4.5 mL), 2x buffer, for use as blank control with Droplet Generation Oil for Probes.