Learn More
Bio Rad Laboratories ddPCR Droplet Reader Oil, 2 L (2 x 1 L), two-color detection system, compatible with QX100 and QX200 Droplet Readers
Catalog No. NC3591196
Encompass
The Bio-Rad Laboratories Inc. ddPCR Droplet Reader Oil is a critical reagent essential for use with QX200 and QX100 Droplet Readers. This oil is specifically designed to facilitate the precise quantification of nucleic acids in Droplet Digital™ PCR workflows by enabling accurate analysis of individual droplets following amplification. It is provided in a 2 L volume, packaged as two 1 L bottles, making it suitable for multiple ddPCR runs.
  • Accurate quantification of PCR-positive and PCR-negative droplets
  • Optimized for seamless integration with both QX100 and QX200 Droplet Readers
  • Supports two-color optical detection systems
  • Fundamental component for reliable droplet analysis in ddPCR workflows
Catalog No. NC3591196 Supplier Bio Rad Laboratories Supplier No. 17005221
The Bio-Rad Laboratories Inc. ddPCR Droplet Reader Oil is a critical reagent essential for use with QX200 and QX100 Droplet Readers. This oil is specifically designed to facilitate the precise quantification of nucleic acids in Droplet Digital™ PCR workflows by enabling accurate analysis of individual droplets following amplification. It is provided in a 2 L volume, packaged as two 1 L bottles, making it suitable for multiple ddPCR runs.
  • Accurate quantification of PCR-positive and PCR-negative droplets
  • Optimized for seamless integration with both QX100 and QX200 Droplet Readers
  • Supports two-color optical detection systems
  • Fundamental component for reliable droplet analysis in ddPCR workflows