Learn More
Bio Rad Laboratories ddPCR Multiplex Supermix, 1.2 ml, 4x concentration, ready-to-use mix, compatible with QX600/QX200 systems, 200 reactions (20 μl each), -20°C storage
Catalog No. NC2754817
Encompass
ddPCR Multiplex Supermix, 1.2 ml, 4x concentration, ready-to-use mix, compatible with QX600/QX200 systems, 200 reactions (20 μl each), -20°C storage
Catalog No. NC2754817 Supplier Bio Rad Laboratories Supplier No. 12005909
ddPCR Multiplex Supermix, 1.2 ml, 4x concentration, ready-to-use mix, compatible with QX600/QX200 systems, 200 reactions (20 μl each), -20°C storage