Learn More
Bio Rad Laboratories ddPCR 96-Well Plates, 25/pk
Catalog No. NC1305904
Encompass
Pkg of 25, clear well/clear shell semi-skirted plates, for use with QX200/QX100 or QX200 AutoDG Droplet Digital PCR Systems.
Catalog No. NC1305904 Supplier Bio Rad Laboratories Supplier No. 12001925
Pkg of 25, clear well/clear shell semi-skirted plates, for use with QX200/QX100 or QX200 AutoDG Droplet Digital PCR Systems.