Learn More
Bio Rad Laboratories ddPCR Supermix for Probes (No dUTP)
Catalog No. NC1305905
Encompass
2 ml (2 x 1 ml), 2x supermix, for use in sample preparation for droplet generation in the QX200/QX100 Droplet Digital PCR Systems.
Catalog No. NC1305905 Supplier Bio Rad Laboratories Supplier No. 1863023
2 ml (2 x 1 ml), 2x supermix, for use in sample preparation for droplet generation in the QX200/QX100 Droplet Digital PCR Systems.