Learn More
Cayman Chemical DDR1-IN-1hydrate 5mg
SDP
Catalog No. 502979795
Encompass_Preferred
A selective DDR1 inhibitor (IC50 = 105 nM; EC50 = 87 nM; inhibits colorectal cancer cell lines when used in combination with the PI3K/mTOR inhibitor, GSK2126458
Catalog No. 50-297-9795 Supplier Cayman Chemical Supplier No. 180925 MG
A selective DDR1 inhibitor (IC50 = 105 nM; EC50 = 87 nM; inhibits colorectal cancer cell lines when used in combination with the PI3K/mTOR inhibitor, GSK2126458