Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DDR1 protein, human (HEK293, His) | >95.0% | 50 UG
SDP

Catalog No. 5000291121
Encompass_Preferred

DDR1 protein is the recombinant extracellular domain of human Discoidin Domain Receptor 1, produced in HEK293 mammalian cells and supplied with a C-terminal His tag as a lyophilized preparation for research use. It is suitable for binding studies, antibody characterization, and assay development, and is provided with purity and handling information in the accompanying documentation.

  • Recombinant extracellular domain of human DDR1 expressed in HEK293 cells.
  • Supplied with a C-terminal His tag for detection or purification.
  • Lyophilized formulation with trehalose for stability during storage.
  • Purity greater than 95% by SDS-PAGE.
  • Molecular weight approximately 55-70 kDa, extracellular domain D21-T416.
  • Store frozen; aliquot and avoid repeated freeze-thaw cycles.

Catalog No. 50-002-91121 Supplier Medchemexpress LLC Supplier No. HYP7530550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DDR1 protein is the recombinant extracellular domain of human Discoidin Domain Receptor 1, produced in HEK293 mammalian cells and supplied with a C-terminal His tag as a lyophilized preparation for research use. It is suitable for binding studies, antibody characterization, and assay development, and is provided with purity and handling information in the accompanying documentation.

  • Recombinant extracellular domain of human DDR1 expressed in HEK293 cells.
  • Supplied with a C-terminal His tag for detection or purification.
  • Lyophilized formulation with trehalose for stability during storage.
  • Purity greater than 95% by SDS-PAGE.
  • Molecular weight approximately 55-70 kDa, extracellular domain D21-T416.
  • Store frozen; aliquot and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.