Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ddr1 protein, mouse (HEK293, C-His, solution) | >95.0% | 10 UG
SDP

Catalog No. 5000291091
Encompass_Preferred

Recombinant mouse DDR1 extracellular domain expressed in HEK293 cells and supplied as a C-terminal His-tagged solution. The product corresponds to amino acids D20-T414, is provided in PBS (0.22 μm filtered), and is intended for research use in studies of collagen receptor signaling and extracellular matrix interactions.

  • Mouse DDR1 extracellular domain, amino acids D20-T414.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag for detection and purification.
  • Supplied as a sterile, 0.22 μm filtered solution in PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Store at -80 °C for long term; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-91091 Supplier Medchemexpress LLC Supplier No. HYP7530210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse DDR1 extracellular domain expressed in HEK293 cells and supplied as a C-terminal His-tagged solution. The product corresponds to amino acids D20-T414, is provided in PBS (0.22 μm filtered), and is intended for research use in studies of collagen receptor signaling and extracellular matrix interactions.

  • Mouse DDR1 extracellular domain, amino acids D20-T414.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag for detection and purification.
  • Supplied as a sterile, 0.22 μm filtered solution in PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Store at -80 °C for long term; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.