Learn More
Medchemexpress LLC DDR1 protein, mouse (HEK293, His) | >95.0% | 100 UG
SDP
Catalog No. 5000292567
Encompass_Preferred
Recombinant mouse discoidin domain receptor 1 (DDR1) protein produced in HEK293 cells and supplied as a 0.22 μm filtered PBS solution with a C-terminal His tag. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 50-60 kDa, and is provided in a 100 μg format for research applications.
  • Expressed in HEK293 for mammalian-like post-translational modifications.
  • Supplied as a 0.22 μm filtered PBS solution ready for assays.
  • C-terminal His tag facilitates purification and detection.
  • High purity (>95%) suitable for biochemical and cell-based studies.
  • Amino acid range corresponds to accession Q03146-2 (D20-T414) representing functional domains.
Catalog No. 50-002-92567 Supplier Medchemexpress LLC Supplier No. HYP75302100UG
Recombinant mouse discoidin domain receptor 1 (DDR1) protein produced in HEK293 cells and supplied as a 0.22 μm filtered PBS solution with a C-terminal His tag. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 50-60 kDa, and is provided in a 100 μg format for research applications.
  • Expressed in HEK293 for mammalian-like post-translational modifications.
  • Supplied as a 0.22 μm filtered PBS solution ready for assays.
  • C-terminal His tag facilitates purification and detection.
  • High purity (>95%) suitable for biochemical and cell-based studies.
  • Amino acid range corresponds to accession Q03146-2 (D20-T414) representing functional domains.