Learn More
Medchemexpress LLC DDR1 protein, mouse (HEK293, His, solution) | 98.6% | 5 UG
SDP
Supplier:  Medchemexpress LLC HYP753025UG
Encompass_Preferred
Recombinant DDR1 protein derived from mouse and expressed in HEK293 mammalian cells. Supplied as a C-terminal His-tagged solution, the product contains the D20-T414 sequence (accession Q03146-2) and is optimized for in vitro biochemical and binding studies investigating receptor-ligand interactions and cell signaling pathways.
  • Expressed in HEK293 mammalian cells.
  • C-terminal His tag for purification and detection.
  • Sequence covers D20-T414 (mouse DDR1).
  • Supplied as a solution suitable for biochemical assays.
  • High purity (>95% by reducing SDS-PAGE).
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-92070