Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DDR1 protein, rat (HEK293, His) | >95.0% | 2 UG
SDP

Catalog No. 5000291119
Encompass_Preferred

Recombinant rat DDR1 protein expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified protein for biochemical assays, receptor-ligand interaction studies, and antibody validation, provided in small research quantities suitable for in vitro experiments.

  • Expression system: HEK293 cells.
  • Tag: C-terminal His tag for purification and detection.
  • Purity: >95% by reducing SDS-PAGE.
  • Applications: biochemical assays, receptor-ligand studies, antibody validation.
  • Available sizes: 2 μg, 5 μg, 10 μg, 50 μg, 100 μg, 500 μg, 1 mg.

Catalog No. 50-002-91119 Supplier Medchemexpress LLC Supplier No. HYP752992UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant rat DDR1 protein expressed in HEK293 cells with a C-terminal His tag. Supplied as a purified protein for biochemical assays, receptor-ligand interaction studies, and antibody validation, provided in small research quantities suitable for in vitro experiments.

  • Expression system: HEK293 cells.
  • Tag: C-terminal His tag for purification and detection.
  • Purity: >95% by reducing SDS-PAGE.
  • Applications: biochemical assays, receptor-ligand studies, antibody validation.
  • Available sizes: 2 μg, 5 μg, 10 μg, 50 μg, 100 μg, 500 μg, 1 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.