Learn More
Medchemexpress LLC DDR2 Human HEK293 5ug | 5ug
SDP
Catalog No. 5000185936
Encompass_Preferred
The DDR2 protein is a tyrosine kinase critical in tissue remodeling acting as a cell surface receptor for fibrillar collagen and affecting cell differentiation extracellular matrix remodeling migration and proliferation DDR2 is critical for bone development and centrally regulates osteoblast differentiation and chondrocyte maturation through MAP kinase signaling and activation of RUNX2 DDR2 Protein Human (HEK293 His) is the recombinant human-derived DDR2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-85936 Supplier Medchemexpress LLC Supplier No. HYP768695UG
The DDR2 protein is a tyrosine kinase critical in tissue remodeling acting as a cell surface receptor for fibrillar collagen and affecting cell differentiation extracellular matrix remodeling migration and proliferation DDR2 is critical for bone development and centrally regulates osteoblast differentiation and chondrocyte maturation through MAP kinase signaling and activation of RUNX2 DDR2 Protein Human (HEK293 His) is the recombinant human-derived DDR2 protein expressed by HEK293 with C-His labeled tag