Learn More
Invitrogen™ Human DDR2, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic Domain, Cytoplasmic
Specifications
Specifications
| Accession Number | Q16832 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 4921 |
| Molecular Weight (g/mol) | 79.8 kDa |
| Name | Human DDR2, GST Tag |
| Quantity | 10 μg |
| Gene Alias | AW495251; CD_antigen: CD167b; CD167 antigen-like family member B; CD167b; CD167b antigen; cell migration-inducing protein 20; Ddr2; Discoidin domain receptor 2; discoidin domain receptor family, member 2; discoidin domain receptor tyrosine kinase 2; discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein; hydroxyaryl-protein kinase; MIG20a; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; neurotrophic tyrosine kinase, receptor-related 3; Ntrkr3; receptor protein-tyrosine kinase TKT; RP11-572K18.1; TKT; TYRO10; tyrosine-protein kinase TYRO 10; tyrosine-protein kinase TYRO10; tyrosylprotein kinase |
| Common Name | DDR2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.