Learn More
Electron Microscopy Sciences DDSA Dodenyl Succinic Anhydride Specially Distilled 450 ML
SDP
Supplier:  Electron Microscopy Sciences 13710
Encompass_Preferred
(Dodecenyl Succinic Anhydride)
Dodecenyl Succinic Anhydride ultra pure is much better than DDSA. It is distilled by a new, slower process, which has a distinct effect on the color control of embedding resins, and offers complete infiltration of tissues. Produces blocks which are totally transparent.
Catalog No. 50-980-377