Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ddtpp (trisodium) solution | 128524-26-5 | 99.9% | 532.11 | 25 MG
SDP

Catalog No. 5000351862
Encompass_Preferred

Ddtpp trisodium solution is a 2',3'-dideoxyribonucleoside 5'-triphosphate (ddNTP) that acts as a chain-elongating inhibitor of DNA polymerase for DNA sequencing. This product is intended for research use only.

  • Purity: 99.94%
  • Molecular weight: 532.11
  • Chemical formula: C10H14N2Na3O13P3
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Application: Used as a chain-elongating inhibitor of DNA polymerase for DNA sequencing
  • Storage: Solution, -20°C, 2 years

Catalog No. 50-003-51862 Supplier Medchemexpress LLC Supplier No. 25MGHY137694B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ddtpp trisodium solution is a 2',3'-dideoxyribonucleoside 5'-triphosphate (ddNTP) that acts as a chain-elongating inhibitor of DNA polymerase for DNA sequencing. This product is intended for research use only.

  • Purity: 99.94%
  • Molecular weight: 532.11
  • Chemical formula: C10H14N2Na3O13P3
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Application: Used as a chain-elongating inhibitor of DNA polymerase for DNA sequencing
  • Storage: Solution, -20°C, 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.